missing translation for 'onlineSavingsMsg'
Learn More

Chromoionophore VI, TRC

Product Code. 30364819
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30364819 50 mg -
30367334 5 mg 5mg
30355423 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30364819 Supplier Toronto Research Chemicals Supplier No. TRCC43293050MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 138833-47-3
Molecular Formula C38H46Br2O5
Molecular Weight (g/mol) 742.59
Synonym 4',5'-Dibromofluorescein Octadecyl Ester, ETH 7075, 2-(4,5-Dibromo-6-hydroxy-3-oxo-3H-xanthen-9-yl)-benzoic Acid Octadecyl Ester
IUPAC Name octadecyl 2-(4,5-dibromo-3-hydroxy-6-oxoxanthen-9-yl)benzoate
SMILES CCCCCCCCCCCCCCCCCCOC(=O)c1ccccc1C2=C3C=CC(=O)C(=C3Oc4c(Br)c(O)ccc24)Br

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C38H46Br2O5/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-19-26-44-38(43)28-21-18-17-20-27(28)33-29-22-24-31(41)34(39)36(29)45-37-30(33)23-25-32(42)35(37)40/h17-18,20-25,41H,2-16,19,26H2,1H3
Formula Weight 740.1712
Chemical Name or Material Chromoionophore VI
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.