missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Chromogranin C Recombinant Rabbit Monoclonal Antibody (HL2992)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA556649
This item is not returnable.
View return policy
Description
Chromogranin C Recombinant Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
The protein encoded by this gene is a member of the chromogranin/secretogranin family of neuroendocrine secretory proteins. Studies in rodents suggest that the full-length protein, secretogranin II, is involved in the packaging or sorting of peptide hormones and neuropeptides into secretory vesicles. The full-length protein is cleaved to produce the active peptide secretoneurin, which exerts chemotaxic effects on specific cell types, and EM66, whose function is unknown.
Specifications
| Chromogranin C | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| P10362, P13521, Q03517 | |
| SCG2 | |
| Recombinant fragment of human Chromogranin C. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| HL2992 | |
| Unconjugated | |
| SCG2 | |
| Chcg; Chgc; Chromogranin C (Secretogranin II); chromogranin-C; EM66; Manserin; SCG2; Scg-2; secretogranin 2; secretogranin II; secretogranin II (chromogranin C); secretogranin-2; Secretoneurin; SgII; SN | |
| Rabbit | |
| Protein A | |
| RUO | |
| 20254, 24765, 7857 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction