missing translation for 'onlineSavingsMsg'
Learn More

Chromogranin C Antibody (SGII 6B1/3) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody

£150.00 - £316.00

Specifications

Antigen Chromogranin C
Clone SGII 6B1/3
Concentration 1 mg/mL
Dilution Western Blot : 1:100 - 1:2000, Flow Cytometry : 1:10 - 1:1000, Immunohistochemistry : 1:10 - 1:500, Immunocytochemistry/Immunofluorescence : 1:10 - 1:500, Immunoprecipitation : 1:10 - 1:500
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18684819
View Documents
Novus Biologicals
NBP2-76658-0.025ml
0.025 mL
£150.00
0.03mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18383881
View Documents
Novus Biologicals
NBP2-76658
0.1 mL
£316.00
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Chromogranin C Monoclonal specifically detects Chromogranin C in Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications

Specifications

Chromogranin C
1 mg/mL
Monoclonal
Mouse
Rat
CHCG, Chromogranin-C, EM66, SCG2, secretogranin IICHGCchromogranin-C, secretoneurin, SgIIsecretogranin-2, SN
SCG2
IgG2b
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
SGII 6B1/3
Western Blot : 1:100 - 1:2000, Flow Cytometry : 1:10 - 1:1000, Immunohistochemistry : 1:10 - 1:500, Immunocytochemistry/Immunofluorescence : 1:10 - 1:500, Immunoprecipitation : 1:10 - 1:500
Unconjugated
Neuroscience
PBS with 0.02% sodium azide
7857
Peptide from rat Chromogranin C (C terminus of p18: KEENSRENPF)
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.