missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chromogranin B Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14968-1-AP-20UL
This item is not returnable.
View return policy
Description
Chromogranin B is a tyrosine sulfated secretory protein found in a wide variety of peptidergic endocrine cells. Chromogranin functions as a neuroendocrine secretory granule protein which may be the precursor for other biologically active peptides.Specifications
| Chromogranin B | |
| Polyclonal | |
| Unconjugated | |
| CHGB | |
| CgB, CHGB, Chromogranin B, SCG1, Secretogranin 1, Secretogranin I, SgI | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 1114, 12653 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P05060, P16014 | |
| CHGB | |
| Chromogranin B Fusion Protein Ag6833 | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction