missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chromogranin B Antibody (CL11393) [Alexa Fluor™ 350], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-21192AF350
This item is not returnable.
View return policy
Description
Chromogranin B Monoclonal antibody specifically detects Chromogranin B in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
Specifications
| Chromogranin B | |
| Monoclonal | |
| Alexa Fluor 350 | |
| CgB, chromogranin B (secretogranin 1), Chromogranin-B, SCG1sgI, secretogranin B, Secretogranin I, secretogranin-1, SgI | |
| This antibody was developed using a recombinant protein derived from P05060, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Cell Biology, Cytokines and Growth Factors, Neuroscience, Signal Transduction | |
| 1114 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| CL11393 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction