missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Chromogranin A Recombinant Mouse Monoclonal Antibody (PD01-39)
GREENER_CHOICE

Product Code. 17919872
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17919872 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17919872 Supplier Invitrogen™ Supplier No. MA550644

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Recombinant Monoclonal Antibody

Positive control: Human pancreas tissue, human thyroid tissue, human atypical carcinoid tissue, human medullary thyroid carcinoma tissue, human appendix tissue. Predicted band size: 51 kDa Subcellular Location: Cytoplasmic vesicle, Secreted.

Chromogranin A (CHGA) is a member of the chromogranin/secretogranin family of neuroendocrine secretory proteins. It is found in secretory vesicles of neurons and endocine cells. CHGA is a precursor to three biologically active peptides: vasostatin, pancreastatin, and parastatin. These peptides act as autocine or paracrine negative modulators of the neuroendocrine system. CHGA is an excellent marker for carcinoid tumors, pheo-chromocytomas, paragangliomas, and other neuro-endocrine tumors. Coexpression of chromogranin A and neuron specific enolase (NSE) is common in neuroendocrine neoplasms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Chromogranin A
Applications Immunohistochemistry (Paraffin)
Classification Recombinant Monoclonal
Clone PD01-39
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene Chga
Gene Accession No. P10645
Gene Alias AL-11; AL26; Beta-granin; betagranin (N-terminal fragment of chromogranin A); catestatin; CgA; CHGA; ChrA; chromofungin; chromogranin A; chromogranin A (parathyroid secretory protein 1); chromogranin A precursor; chromogranin-A; CTNNB; DKFZp686D02253; EA-92; ER-37; ES-43; FLJ25606; FLJ37923; GE-25; GR-44; GV-19; LF-19; LOW QUALITY PROTEIN: chromogranin-A; MGC105435; Pancreastatin; Parastatin; parathyroid secretory protein 1; p-Glu serpinin precursor; pituitary secretory protein I; Serpinin; Serpinin-RRG; SL21; SP-I; SS-18; Vasostatin I; Vasostatin II; Vasostatin-1; Vasostatin-2; WA-8; WE-14
Gene Symbols Chga
Host Species Mouse
Immunogen Recombinant full length protein corresponding to Human Chromogranin A.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1113
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.