missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chromogranin A Mouse anti-Human, Mouse, Pig, Rat, Clone: 2A7D11, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 60135-2-IG-150UL
This item is not returnable.
View return policy
Description
Chromogranin A Monoclonal antibody specifically detects Chromogranin A in Human, Mouse, Pig, Rat samples. It is validated for IHC, WB,ELISASpecifications
| Chromogranin A | |
| Monoclonal | |
| 1000 μg/mL | |
| PBS with 0.02% sodium azide and 50% glycerol pH 7.3. | |
| Mouse | |
| 150 μL | |
| 1113 | |
| Store at -20°C | |
| Liquid |
| Immunohistochemistry, Western Blot, ELISA | |
| 2A7D11 | |
| Unconjugated | |
| BC024028 | |
| Protein G purification | |
| Primary | |
| Human, Mouse, Pig, Rat | |
| Antibody | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction