Learn More
Invitrogen™ Chromogranin A Monoclonal Antibody (1A6B6)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA517056
Description
MA5-17056 targets CHGA in indirect ELISA and WB applications and shows reactivity with Human and Mouse samples. The MA5-17056 immunogen is purified recombinant fragment of human CHGA (amino acids: 87-252 ) expressed in E. Coli. MA5-17056 detects CHGA which has a predicted molecular weight of approximately 50.7kDa.
Chromogranin A (CHGA) is a member of the chromogranin/secretogranin family of neuroendocrine secretory proteins. It is found in secretory vesicles of neurons and endocine cells. CHGA is a precursor to three biologically active peptides: vasostatin, pancreastatin, and parastatin. These peptides act as autocine or paracrine negative modulators of the neuroendocrine system. CHGA is an excellent marker for carcinoid tumors, pheo-chromocytomas, paragangliomas, and other neuro-endocrine tumors. Coexpression of chromogranin A and neuron specific enolase (NSE) is common in neuroendocrine neoplasms.
Specifications
| Chromogranin A | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.05% sodium azide | |
| P10354, P10645, P26339 | |
| Chga | |
| Purified recombinant fragment of human CHGA (amino acids: 87-252 ) expressed in E. Coli. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
| ELISA, Western Blot | |
| 1A6B6 | |
| Unconjugated | |
| Chga | |
| AL-11; AL26; Beta-granin; betagranin (N-terminal fragment of chromogranin A); catestatin; CgA; CHGA; ChrA; chromofungin; chromogranin A; chromogranin A (parathyroid secretory protein 1); chromogranin A precursor; chromogranin-A; CTNNB; DKFZp686D02253; EA-92; ER-37; ES-43; FLJ25606; FLJ37923; GE-25; GR-44; GV-19; LF-19; LOW QUALITY PROTEIN: chromogranin-A; MGC105435; Pancreastatin; Parastatin; parathyroid secretory protein 1; p-Glu serpinin precursor; pituitary secretory protein I; Serpinin; Serpinin-RRG; SL21; SP-I; SS-18; Vasostatin I; Vasostatin II; Vasostatin-1; Vasostatin-2; WA-8; WE-14 | |
| Mouse | |
| Protein G | |
| RUO | |
| 1113, 12652, 24258 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.