missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chromogranin A Antibody (CHGA/1731R), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-53139-100UG
This item is not returnable.
View return policy
Description
Chromogranin A Monoclonal specifically detects Chromogranin A in Human, Rat (Negative) samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array.
Specifications
| Chromogranin A | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, Immunohistochemistry-Paraffin 0.25 - 0.5 ug/ml | |
| betagranin (N-terminal fragment of chromogranin A), CGA, chromogranin A (parathyroid secretory protein 1), chromogranin-A, parathyroid secretory protein 1, Pituitary secretory protein I, SP-I | |
| Rabbit | |
| 72 kDa | |
| 100 μg | |
| Apoptosis, Cancer, Neuronal Cell Markers, Tumor Biomarkers | |
| 1113 | |
| Store at 4C. | |
| IgG κ |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| CHGA/1731R | |
| Unconjugated | |
| 10mM PBS with 0.05% BSA with 0.05% Sodium Azide | |
| CHGA | |
| Recombinant human full-length Chromogranin A protein | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Rat (Negative) | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction