missing translation for 'onlineSavingsMsg'
Learn More

Chromium (VI) Oxide, TRC

Product Code. 30360458
Change view
Click to view available options
Quantity:
10 g
100 g
Unit Size:
100g
10g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30360458 100 g 100g
30356801 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30360458 Supplier Toronto Research Chemicals Supplier No. TRCC432770100G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1333-82-0
Molecular Formula Cr O3
Molecular Weight (g/mol) 99.99
Synonym Chromia, Chromic Anhydride, Chromic Trioxide, Chromium Oxide, Chromium Trioxide, Chromium(VI) Trioxide, Monochromium Trioxide
IUPAC Name trioxochromium
SMILES O=[Cr](=O)=O

Specifications

Quantity 100 g
InChI Formula InChI=1S/Cr.3O
Formula Weight 99.9253
Chemical Name or Material Chromium (VI) oxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.