missing translation for 'onlineSavingsMsg'
Learn More

Chromium(VI) oxide, 99%, Thermo Scientific Chemicals

Catalog Number 11381849
Click to view available options
Quantity:
250 g
1 kg
5 x 1 kg
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
1333-82-0
CrO3
99.99
MFCD00010952
WGLPBDUCMAPZCE-UHFFFAOYSA-N
chromium trioxide, chromium vi oxide, monochromium trioxide, chromtrioxid, chromic trioxide, chromsaeureanhydrid, anhydride chromique, chromium 6+ oxide, chromium vi trioxide, chromium oxide
14915
CHEBI:48240
trioxochromium
O=[Cr](=O)=O
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
1333-82-0
CrO3
99.99
MFCD00010952
WGLPBDUCMAPZCE-UHFFFAOYSA-N
chromium trioxide, chromium vi oxide, monochromium trioxide, chromtrioxid, chromic trioxide, chromsaeureanhydrid, anhydride chromique, chromium 6+ oxide, chromium vi trioxide, chromium oxide
14915
CHEBI:48240
trioxochromium
O=[Cr](=O)=O

It is widely used in chromium electroplating, and as protective coatings in corrosion/oxidation resistance of metals. It is also employed in CDs and DVDs. It is useful as an oxidant in Jones oxidation in acetic acid or acetone. It is used in the oxidation of primary alcohols into carboxylic acids, and secondary alcohols into ketones. With phosphoric acid, it is used as a stripping agent for anodic coatings of all types and in the production of syntheric rubies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 250°C (decomposition)
Melting Point 196°C
Physical Form Crystalline
Quantity 1 kg
Assay Percent Range 99%
Solubility Information Very soluble in water. Soluble in sulfuric,nitric acid,acetic acid,and acetone.
Formula Weight 99.99
Sensitivity Hygroscopic
Appearance Dark red crystals
Density 2.7
Chemical Name or Material Chromium(VI) oxide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'