missing translation for 'onlineSavingsMsg'
Learn More

Chromium sputtering target, 50.8mm (2.0 in.) dia. x 6.35mm (0.250 in.) thick, 99.95% (metals basis), Thermo Scientific™

Product Code. 15484785
Click to view available options
Quantity:
1 Ea.
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 7440-47-3
Molecular Formula Cr
MDL Number MFCD00010944
Quantity 1 Ea.
Synonym chrom, chrome, cromo, metal, chrome french, compounds, chrom german, chromium, elemental, chromium, metal, ccris 159
Solubility Information Insoluble in water.
InChI Key VYZAMTAEIAYCRO-UHFFFAOYSA-N
SMILES [Cr]
IUPAC Name chromium
Molecular Weight (g/mol) 52.00
PubChem CID 23976
ChEBI CHEBI:28073
Percent Purity 99.95%
Odor Odorless
Assay (metals basis)
Chemical Name or Material Chromium sputtering target, 50.8mm (2.0 in.) dia. x 6.35mm (0.250 in.) thick
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'