missing translation for 'onlineSavingsMsg'
Learn More

Chromium silicide, 99+% (metals basis)

Product Code. 11336528
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11336528 50 g 50g
11326528 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11336528 Supplier Thermo Scientific Alfa Aesar Supplier No. 012151.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12018-09-6 | CrSi2 | 108.166 g/mol

Chromium silicide is applied in production of ceramic materials. It can also produce an oxidation-resistant surface in a low pressure plasma spraying.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12018-09-6
Molecular Formula CrSi2
Molecular Weight (g/mol) 108.166
MDL Number MFCD00168070
InChI Key CHXGWONBPAADHP-UHFFFAOYSA-N
Synonym chromsilicid, chromium silicide, bis $l^ 3
PubChem CID 45051936
IUPAC Name bis($l^{3}-silanylidyne)chromium
SMILES [Si]#[Cr]#[Si]

Specifications

Melting Point 1490°C
Color Gray
Physical Form Powder
Quantity 50 g
Assay Percent Range >95%
Solubility Information Insoluble in water.
Formula Weight 108.17
Odor Odorless
Sensitivity Moisture sensitive
Appearance Gray powder
Chemical Name or Material Chromium silicide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.