missing translation for 'onlineSavingsMsg'
Learn More

Chromium powder, -100 mesh, 99% (metals basis), Thermo Scientific Chemicals

Catalog Number 11391198
Click to view available options
Quantity:
10 g
100 g
500 g
2 kg
This item is not returnable. View return policy
7440-47-3
Cr
52.00
MFCD00010944
VYZAMTAEIAYCRO-UHFFFAOYSA-N
chrom, chrome, cromo, metal, chrome french, compounds, chrom german, chromium, elemental, chromium, metal, ccris 159
23976
CHEBI:28073
chromium
[Cr]
This item is not returnable. View return policy
7440-47-3
Cr
52.00
MFCD00010944
VYZAMTAEIAYCRO-UHFFFAOYSA-N
chrom, chrome, cromo, metal, chrome french, compounds, chrom german, chromium, elemental, chromium, metal, ccris 159
23976
CHEBI:28073
chromium
[Cr]

Apparent density 2.0-3.0g/cm3

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Gray
Appearance Gray powder
Assay Percent Range 99% (Metals basis)
Quantity 2 kg
Mesh Size -100 Mesh
Odor Odorless
Form Powder
Chemical Name or Material Chromium

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'