missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chromium nitride, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
Description
Chromium nitride is used for stainless steel, corrosion resistance steel, alloy steel and such special steel.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Physical Form | Powder |
Quantity | 25 g |
Solubility Information | Insoluble in water. |
Chemical Name or Material | Chromium nitride |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'