missing translation for 'onlineSavingsMsg'
Learn More

Chromium(III) Potassium Sulfate Dodecahydrate, purum p.a, ≥98.5% (RT), Honeywell™

Product Code. p-7120328 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
100 g
1 kg
Packaging:
Poly bottle
Unit Size:
100g
1kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15625930 100 g Poly bottle 100g
15635930 1 kg Poly bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15625930 Supplier Honeywell Research Chemicals Supplier No. 60152100G
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Purum p.a.,≥98.5% (RT)

  • Products with a guarantee certificate and/or suitable for the stated analytical application.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7788-99-0
Molecular Formula CrH24KO20S2
Molecular Weight (g/mol) 499.386
MDL Number MFCD00149917
InChI Key ZFVHBEKVAITXHW-UHFFFAOYSA-J
Synonym chromium potassium sulfate dodecahydrate, chrome alum dodecahydrate, potassium chromium alum dodecahydrate, unii-h54d055wx6, ccris 8929, chromic potassium sulfate dodecahydrate, chromium iii potassium sulfate 1:1:2 , dodecahydrate, sulfuric acid, chromium 3+ potassium salt 2:1:1 , dodecahydrate, acmc-20alga, ksc492e4p
PubChem CID 24596
IUPAC Name potassium;chromium(3+);disulfate;dodecahydrate
SMILES O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[K+].[Cr+3]

Specifications

Melting Point 89°C
Quantity 100 g
Linear Formula CrK(SO4)2 · 12H2O
UN Number NONH for all modes of transport
Formula Weight 499.4g/mol
Percent Purity ≥98.5% (RT)
Packaging Poly bottle
Density 1.830g/cm3
Chemical Name or Material Chromium(III) potassium sulfate dodecahydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.