Learn More
Chromium(III) potassium sulfate dodecahydrate, 98%
Description
Chromium(III) potassium sulfate is used for tanning of leather and as a photographic film hardener. It is used in manufacturing of chromium-containing catalyst.It is used in mordantin in printing and dyeing industry and it could also be used as analytical reagent. In addition, it is also used as food additive.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chromium(III) potassium sulfate is used for tanning of leather and as a photographic film hardener. It is used in manufacturing of chromium-containing catalyst.It is used in mordantin in printing and dyeing industry and it could also be used as analytical reagent. In addition, it is also used as food additive.
Solubility
Soluble in water. 24 g/100 mL (15°C)
Notes
Incompatible with magnesium, aluminum. Store away from heat.
Specifications
Specifications
| Melting Point | 89°C |
| Quantity | 2500 g |
| Linear Formula | CrK(SO4)2·12H2O |
| Merck Index | 14,2229 |
| Solubility Information | Soluble in water. 24g/100 mL (15°C) |
| Formula Weight | 499.39 (283.21 Anhydrous) |
| Percent Purity | 98% |
| Odor | Odorless |
| Density | 1.83 |
| Chemical Name or Material | Chromium (III) potassium sulfate dodecahydrate |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.