missing translation for 'onlineSavingsMsg'
Learn More

Chromium(III) phosphate tetrahydrate

Product Code. 11372648
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11372648 10 g 10g
11382648 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11372648 Supplier Thermo Scientific Alfa Aesar Supplier No. 024110.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For Use As a Catalyst, Pigment | CAS: 10101-59-4 | CrH8O8P | 219.026 g/mol

It is used as an additive in drugs, anti-corrosive pigments, catalysts and polymer manufacture, is also used to catalyze cation exchange in sorption reactions, as a laminated structure to dampen vibration and noise in a motor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10101-59-4
Molecular Formula CrH8O8P
Molecular Weight (g/mol) 219.026
MDL Number MFCD00016049
InChI Key BVZHAJJRIRDSTG-UHFFFAOYSA-K
Synonym chromium iii phosphate tetrahydrate, cr.po4.4h2o, chromium 3+ ion tetrahydrate phosphate, chromium 3+ phosphate-water 1/1/4, chromium 3+ tetrahydrate phosphate
PubChem CID 53471880
IUPAC Name chromium(3+);phosphate;tetrahydrate
SMILES O.O.O.O.[O-]P(=O)([O-])[O-].[Cr+3]

Specifications

Physical Form Powder
Quantity 10 g
Solubility Information Soluble in acids. Insoluble in water.
Formula Weight 219.04 (146.97 Anhydrous)
Appearance Dark powder
Density 2.26 g/cm3
Chemical Name or Material Chromium(III) phosphate tetrahydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.