missing translation for 'onlineSavingsMsg'
Learn More

Chromium(III) oxide, Puratronic™, 99.97% (metals basis)

Product Code. 11330939
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Unit Size:
100g
25g
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11330939 500 g 500g
11340939 5 g 5g
11350939 25 g 25g
11360939 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11330939 Supplier Thermo Scientific Alfa Aesar Supplier No. 010688.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1308-38-9 | Cr2O3 | 151.99 g/mol

Also known as green compound for buffing and polishing, it can be used as abrasives. Apart from being used as a precursor to the magnetic pigment viz., chromium dioxide, it is used in inks, glasses, and pigments (viridian). They are employed as plating agents and surface treating agents. It is a catalyst in the Phillips process of manufacture of high density polythene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1308-38-9
Molecular Formula Cr2O3
Molecular Weight (g/mol) 151.99
MDL Number MFCD00010949
InChI Key UOUJSJZBMCDAEU-UHFFFAOYSA-N
Synonym chromium iii oxide, chromia, dichromium trioxide, chromium sesquioxide, chrome green, green chromium oxide, chromium 3+ oxide, green cinnabar, chrome ochre, chrome oxide
PubChem CID 517277
ChEBI CHEBI:48242
SMILES [O--].[O--].[O--].[Cr+3].[Cr+3]

Specifications

Boiling Point 3000°C
Melting Point 2435°C
Color Green
Physical Form Powder
Quantity 500 g
Merck Index 14,2234
Solubility Information Insoluble in water,alcohol,acetone,alkalies and acids.
IUPAC Name oxo(oxochromiooxy)chromium
Formula Weight 151.99
Percent Purity 99.97%
Grade Puratronic
Odor Odorless
Refractive Index 2.551
Density 5.22 g/mL
Assay (metals basis)
Chemical Name or Material Chromium(III) oxide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.