missing translation for 'onlineSavingsMsg'
Learn More

Chromium(III) oxide, 99%, Thermo Scientific Chemicals

Catalog Number 11381629
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
1308-38-9
Cr2O3
151.99
MFCD00010949
UOUJSJZBMCDAEU-UHFFFAOYSA-N
chromium iii oxide, chromia, dichromium trioxide, chromium sesquioxide, chrome green, green chromium oxide, chromium 3+ oxide, green cinnabar, chrome ochre, chrome oxide
517277
CHEBI:48242
dichromium(3+) trioxidandiide
[O--].[O--].[O--].[Cr+3].[Cr+3]
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
1308-38-9
Cr2O3
151.99
MFCD00010949
UOUJSJZBMCDAEU-UHFFFAOYSA-N
chromium iii oxide, chromia, dichromium trioxide, chromium sesquioxide, chrome green, green chromium oxide, chromium 3+ oxide, green cinnabar, chrome ochre, chrome oxide
517277
CHEBI:48242
dichromium(3+) trioxidandiide
[O--].[O--].[O--].[Cr+3].[Cr+3]

Chromium(III) oxide is known as green compound which is used for buffing and polishing. It can be used as abrasives. Apart from being used as a precursor to the magnetic pigment viz., chromium dioxide, it is used in inks, glasses, and pigments (viridian). They are employed as plating agents and surface treating agents. It is a catalyst in the Phillips process of manufacture of high density polythene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 3000°C
Melting Point 2435°C
Color Green
Physical Form Powder
Quantity 500 g
Assay Percent Range 99%
Solubility Information Insoluble in water,alcohol,acetone and acids.
Formula Weight 151.99
Odor Odorless
Refractive Index 2.551
Appearance Pale to dark green powder
Density 5.22
Chemical Name or Material Chromium(III) oxide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'