missing translation for 'onlineSavingsMsg'
Learn More

Chromium(Iii) Oxide, 99.9% trace metals basis, Honeywell™

Product Code. 15691740 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Poly Bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15691740 25 g Poly Bottle 25g
15601750 5 g Poly Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15691740 Supplier Honeywell Research Chemicals Supplier No. 20306825G
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Powder, 99.9% trace metals basis

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
    Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
    Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1308-38-9
Molecular Formula Cr2O3
Molecular Weight (g/mol) 151.99
MDL Number MFCD00010949
InChI Key UOUJSJZBMCDAEU-UHFFFAOYSA-N
Synonym chromium iii oxide, chromia, dichromium trioxide, chromium sesquioxide, chrome green, green chromium oxide, chromium 3+ oxide, green cinnabar, chrome ochre, chrome oxide
PubChem CID 517277
ChEBI CHEBI:48242
SMILES [O--].[O--].[O--].[Cr+3].[Cr+3]

Specifications

Boiling Point 4000°C
Melting Point 2435°C
Color Green
Physical Form Powder
Quantity 25 g
UN Number NONH for all modes of transport
IUPAC Name dichromium(3+) trioxidandiide
Formula Weight 151.99g/mol
Percent Purity 99.9% (trace metals basis)
Packaging Poly Bottle
Density 5.2g/cu.cm
Chemical Name or Material Chromium(Iii) Oxide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.