missing translation for 'onlineSavingsMsg'
Learn More

Chromium(Iii) Nitrate Nonahydrate, 99%, Honeywell Fluka™

Product Code. p-7120323 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
100 g
500 g
2 kg
Packaging:
Poly Bottle
Unit Size:
100g
2kg
500g
Product Code. Quantity Packaging unitSize
15642430 100 g Poly Bottle 100g
15662430 500 g Poly Bottle 500g
15652430 2 kg Poly Bottle 2kg
3 options
This item is not returnable. View return policy

Product Code. 15642430

Brand: Honeywell-Fluka 239259100G

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

≥99%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-02-8
Molecular Formula CrN3O9
Molecular Weight (g/mol) 238.01
MDL Number MFCD00149659
InChI Key PHFQLYPOURZARY-UHFFFAOYSA-N
Synonym chromium iii nitrate nonahydrate, unii-d2806iol1l, nitric acid, chromium 3+ salt, nonahydrate, acmc-20alg5, bis 7-chlorophenyl sulfone, cr.3no3.9h2o, ksc495a9j, chromium trinitrate, nonahydrate, chromium iii nitrate, nonahydrate, chromium 3+ nonahydrate trinitronate
PubChem CID 9887206
IUPAC Name chromium(3+);trinitrate;nonahydrate
SMILES [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Cr+3]

Specifications

Melting Point 60°C
Color Black to Blue
Quantity 100 g
Linear Formula Cr(NO3)3 · 9H2O
UN Number UN2720
Formula Weight 400.15g/mol
Percent Purity 99%
Packaging Poly Bottle
Appearance Crystals or Chunk(s)
Density 1.89g/cm3
Chemical Name or Material Chromium(Iii) Nitrate Nonahydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.