missing translation for 'onlineSavingsMsg'
Learn More

Chromium(III) nitrate nonahydrate, 99.99% min (metals basis)

Product Code. 11394868
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11394868 100 g 100g
11304878 5 g 5g
11314878 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11394868 Supplier Thermo Scientific Alfa Aesar Supplier No. 043182.22
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7789-02-8 | CrN3O9 | 238.01 g/mol

Chromium nitrate is used in the manufacture of alkali metal-free catalysts and in preservation. It is used in the preparation of chrome catalysts, in textile printing operations, and as a corrosion inhibitor. Chromium(III) nitrate as a source of chromium is added to food supplements for nutritional purposes .

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-02-8
Molecular Formula CrN3O9
Molecular Weight (g/mol) 238.01
MDL Number MFCD00149659
InChI Key PHFQLYPOURZARY-UHFFFAOYSA-N
Synonym chromium iii nitrate nonahydrate, unii-d2806iol1l, nitric acid, chromium 3+ salt, nonahydrate, acmc-20alg5, bis 7-chlorophenyl sulfone, cr.3no3.9h2o, ksc495a9j, chromium trinitrate, nonahydrate, chromium iii nitrate, nonahydrate, chromium 3+ nonahydrate trinitronate
PubChem CID 9887206
IUPAC Name chromium(3+);trinitrate;nonahydrate
SMILES [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Cr+3]

Specifications

Melting Point 60°C
Color Purple
Physical Form Crystalline
Quantity 100 g
Odor Odorless
UN Number UN2720
Merck Index 14,2226
Solubility Information Soluble in water and alcohol.
Formula Weight 400.14 (238.01 Anhydrous)
Percent Purity ≥99.99%
Assay (metals basis)
Chemical Name or Material Chromium (III) nitrate nonahydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.