missing translation for 'onlineSavingsMsg'
Learn More

Chromium(III) Nitrate Monohydrate, Extra Pure, SLR, Fisher Chemical

Product Code. 10704371 Shop All Fisher Chemical Products
Change view
Click to view available options
500g, Glass Jar
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10704371 500g, Glass Jar
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10704371 Supplier Fisher Chemical Supplier No. C/5560/53
Quantity Price
1 £334.93 / 500g
4 £276.48 / 500g

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Melting Point 60°C
CAS 7789-02-8
Color Black
pH 2.0 to 3.0
Physical Form Solid
Quantity 500 g
Molecular Formula CrN3O9
MDL Number 149659
InChI Key PHFQLYPOURZARY-UHFFFAOYSA-N
Molecular Weight (g/mol) 238.01
Formula Weight 400.14g/mol
Packaging Glass Jar
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Chromium(III) nitrate nonahydrate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Oxidising solid Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H272-May intensify fire; oxidiser.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.