missing translation for 'onlineSavingsMsg'
Learn More

Chromium(III) fluoride hydrate, Thermo Scientific™

Product Code. 11341709
Change view
Click to view available options
Quantity:
250 g
1 kg
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11341709 1 kg 1kg
11351709 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11341709 Supplier Thermo Scientific Alfa Aesar Supplier No. 012339.A1
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123333-98-2
Molecular Formula CrF3
Molecular Weight (g/mol) 108.99
MDL Number MFCD00016045
InChI Key FTBATIJJKIIOTP-UHFFFAOYSA-K
IUPAC Name chromium(3+) trifluoride
SMILES [F-].[F-].[F-].[Cr+3]

Specifications

Melting Point >1000°C
Color Green
Physical Form Crystalline
Quantity 1 kg
UN Number UN1756
Merck Index 14,2223
Solubility Information Insoluble in water.
Formula Weight 108.99 (Anhydrous)
Sensitivity Hygroscopic
Chemical Name or Material Chromium(III) fluoride hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.