missing translation for 'onlineSavingsMsg'
Learn More

Chromium(III) fluoride, 98%

Product Code. 11358617
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
This item is not returnable. View return policy

Product Code. 11358617

Brand: Thermo Scientific Alfa Aesar 035741.04

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7788-97-8

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7788-97-8
MDL Number MFCD00016045
Synonym chromium fluoride, chromic fluoride, chromium 3+ trifluoride, acmc-1bht2, chromium 3+ ion trifluoride, chromic fluoride, solid un1756 corrosive, chromic fluoride, solution un1757 corrosive

Specifications

Melting Point >1000°C
Color Green
Physical Form Powder
Quantity 2 g
UN Number UN1756
Merck Index 14,2223
Formula Weight 108.99
Percent Purity 99.98%
Odor Odorless
Sensitivity Hygroscopic
Density 3.8 g/mL
Assay (metals basis)
Chemical Name or Material Chromium(III) fluoride
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.