missing translation for 'onlineSavingsMsg'
Learn More

Chromium(III) chloride, anhydrous, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog Number 11393867
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
10025-73-7
CrCl3
MFCD00010948
chromium iii chloride, chromium 3+ trichloride, anhydrous chromium iii chloride, basic chromic chloride, chromium chloride, basic, chromium trichl, chromium iii ; chloride, acmc-1ai6u, ksc492k0f
This item is not returnable. View return policy
10025-73-7
CrCl3
MFCD00010948
chromium iii chloride, chromium 3+ trichloride, anhydrous chromium iii chloride, basic chromic chloride, chromium chloride, basic, chromium trichl, chromium iii ; chloride, acmc-1ai6u, ksc492k0f

The vapor-phase co-reductions with other metal halides by hydrogen results in finely divided intermetallics with applications as structural materials or compounds with useful thermoelectric, magnetic, and oxidation-resistance properties. It is used as a catalyst, for organic synthesis, for manufacturing dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point ∽1150°C
Color Violet
Physical Form Flakes
Quantity 25 g
Assay Percent Range 99.9% (Metals basis)
Solubility Information Soluble in alcohol,water.
Formula Weight 158.35
Odor Odorless
Sensitivity Hygroscopic
Density 2.87
Chemical Name or Material Chromium(III) chloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'