missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chromium(III) Chloride Hexahydrate, TRC
Click to view available options
Quantity:
250 g
Unit Size:
250g
Specifications
Specifications
| Chemical Name or Material | Chromium(III) Chloride Hexahydrate |
| CAS | 10060-12-5 |
| Molecular Formula | Cl3CrH12O6 |
| Quantity | 250 g |
| Synonym | Chromium chloride hexahydrate, Chromic chloride hexahydrate, Chromium chloride hexahydrate, Chromium trichloride hexahydrate, Chromium(III) chloride hexahydrate, Hexaaquachromium chloride, Hexaaquachromium(III) chloride |
| InChI Formula | InChI=1S/3ClH.Cr.6H2O/h3*1H;;6*1H2/q;;;+3;;;;;;/p-3 |
| SMILES | O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Cr+3] |
| IUPAC Name | chromium(3+);trichloride;hexahydrate |
| Molecular Weight (g/mol) | 266.45 |
| Formula Weight | 264.9105 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction