missing translation for 'onlineSavingsMsg'
Learn More

Chromium(III) chloride hexahydrate, Puratronic™, 99.998% (metals basis)

Product Code. 11300939
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11300939

Brand: Thermo Scientific Alfa Aesar 010686.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10060-12-5 | Cl3CrH12O6 | 266.44 g/mol

Chromium(III) chloride hexahydrate is used as an organic synthesis catalyst and as an additive to intravenous solutions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

 
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10060-12-5
Molecular Formula Cl3CrH12O6
Molecular Weight (g/mol) 266.44
MDL Number MFCD00149660
InChI Key LJAOOBNHPFKCDR-UHFFFAOYSA-K
Synonym chromium iii chloride hexahydrate, chromic chloride hexahydrate, hexaaquachromium chloride, chromium trichloride hexahydrate, trichlorochromium hexahydrate, chromium chloride hexahydrate, hexaaquachromium iii chloride, chrom iii-chlorid, chromium 3+ chloride hexahydrate, chlorid chromity hexahydrat czech
PubChem CID 104957
ChEBI CHEBI:53442
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Cr+3]

Specifications

Melting Point 83°C
Color Green
Physical Form Crystalline Aggregates
Quantity 25 g
Linear Formula CrCl3·6H2O
UN Number UN3260
Merck Index 14,2222
Solubility Information Soluble in water and alcohol. Insoluble in diethyl ether.
IUPAC Name trichlorochromium;hexahydrate
Formula Weight 266.45 (158.35 Anhydrous)
Percent Purity 99.998%
Grade Puratronic
Odor Odorless
Density 1.835 g/mL
Assay (metals basis)
Chemical Name or Material Chromium(III) chloride hexahydrate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.