missing translation for 'onlineSavingsMsg'
Learn More

Chromium(III) Chloride Hexahydrate, MP Biomedicals

Product Code. 11480582 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11480582 500 g 500g
11470582 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11480582 Supplier MP Biomedicals Supplier No. 0219403390
 Restricted Item
Estimated Shipment: 22-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 10060-12-5 | Cl3CrH12O6 | 266.44 g/mol

Chemical Identifiers

CAS 10060-12-5
Molecular Formula Cl3CrH12O6
Molecular Weight (g/mol) 266.44
MDL Number MFCD00149660
InChI Key LJAOOBNHPFKCDR-UHFFFAOYSA-K
Synonym chromium iii chloride hexahydrate, chromic chloride hexahydrate, hexaaquachromium chloride, chromium trichloride hexahydrate, trichlorochromium hexahydrate, chromium chloride hexahydrate, hexaaquachromium iii chloride, chrom iii-chlorid, chromium 3+ chloride hexahydrate, chlorid chromity hexahydrat czech
PubChem CID 104957
ChEBI CHEBI:53442
IUPAC Name chromium(3+) hexahydrate trichloride
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Cr+3]

Specifications

Physical Form Powder
Quantity 500 g
Formula Weight 266.4
Percent Purity 99.5%
Chemical Name or Material Chromium(III) Chloride Hexahydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.