missing translation for 'onlineSavingsMsg'
Learn More

Chromium(III) chloride hexahydrate, 98%

Product Code. 11309777
Change view
Click to view available options
Quantity:
250 g
1 kg
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11309777 250 g 250g
11399767 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11309777 Supplier Thermo Scientific Alfa Aesar Supplier No. 042113.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10060-12-5 | Cl3CrH12O6 | 266.44 g/mol

Chromium(III) chloride hexahydrate is used as a catalyst, as a mordant in printing and dyeing industry. It is involved in the preparation of anhydrous tetrahydrofuran complex of chromium(III) chloride by reacting with trimethylsilylchloride. It finds application in the manufacturing of a variety of chromium-containing pigment in Paint industry and other chromium salts in the chemical industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10060-12-5
Molecular Formula Cl3CrH12O6
Molecular Weight (g/mol) 266.44
MDL Number MFCD00149660
InChI Key LJAOOBNHPFKCDR-UHFFFAOYSA-K
Synonym chromium iii chloride hexahydrate, chromic chloride hexahydrate, hexaaquachromium chloride, chromium trichloride hexahydrate, trichlorochromium hexahydrate, chromium chloride hexahydrate, hexaaquachromium iii chloride, chrom iii-chlorid, chromium 3+ chloride hexahydrate, chlorid chromity hexahydrat czech
PubChem CID 104957
ChEBI CHEBI:53442
IUPAC Name chromium(3+) hexahydrate trichloride
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Cr+3]

Specifications

Melting Point 83°C
Color Green
Physical Form Crystalline
Quantity 250 g
UN Number UN3260
Merck Index 14,2222
Solubility Information Slightly soluble in acetone; Soluble in water, alcohol; Insoluble in ether
Formula Weight 266.45 (158.35 Anhydrous)
Percent Purity 98%
Odor Odorless
Density 1.835 g/mL
Chemical Name or Material Chromium(III) chloride hexahydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.