missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chromium(III) chloride hexahydrate, 98%, Thermo Scientific Chemicals
36.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Use promo code "22914" to get your promotional price.
Click to view available options
Quantity:
500 g
5 kg
Packaging:
Plastic bottle
Plastic drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 83.0°C |
Color | Green |
Physical Form | Crystalline Powder |
Quantity | 500 g |
Assay Percent Range | 98% |
Linear Formula | [Cr(H2O)4Cl2]Cl·2H2O |
Merck Index | 14, 2222 |
Solubility Information | Solubility in water: soluble. Other solubilities: soluble in alcohol, slightly soluble in acetone, practically insoluble in ether |
Formula Weight | 266.44 |
Percent Purity | 98% |
Show More |
Safety and Handling
Product Identifier
- Chromium(III) chloride hexahydrate
- Danger
- Acute toxicity Category 4
- Skin corrosion/irritation Category 1 C
- Serious eye damage/eye irritation Category 1
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : Chromium (III) Chloride Hexahydrate, Chromic chloride
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'