missing translation for 'onlineSavingsMsg'
Learn More

Chromium(III) chloride, anhydrous, 98%, Thermo Scientific Chemicals

Catalog Number 11351699
Click to view available options
Quantity:
50 g
250 g
10025-73-7
MFCD00010948
chromium iii chloride, chromium 3+ trichloride, anhydrous chromium iii chloride, basic chromic chloride, chromium chloride, basic, chromium trichl, chromium iii ; chloride, acmc-1ai6u, ksc492k0f
10025-73-7
MFCD00010948
chromium iii chloride, chromium 3+ trichloride, anhydrous chromium iii chloride, basic chromic chloride, chromium chloride, basic, chromium trichl, chromium iii ; chloride, acmc-1ai6u, ksc492k0f

Chromium chloride and chromium sulfate have been used as dietary supplements. Chromium(III) chloride and chromium(III) sulfate can be used in the manufacture of food for particular nutritional uses and in food supplements. It is found in miscellaneous applications, such as catalysts, hardeners, mordents, crosslinking and binding agents. It is used to make very pure chromium and other chromium compounds, as a mordant in dyeing cotton, and in chromizing metals. It is too used for trivalent electroplating .

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 1300°C (sublimation)
Color Violet
Physical Form Crystalline
Quantity 50 g
UN Number UN3260
Merck Index 14,2222
Solubility Information Partially soluble in water.
Formula Weight 158.35
Percent Purity 98%
Sensitivity Hygroscopic
Density 2.87 g/mL
Chemical Name or Material Chromium(III) chloride, Anhydrous
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'