Learn More
Chromium(III) chloride, anhydrous, 98%, Thermo Scientific Chemicals
Use promo code "21615" to get your promotional price.
Description
Chromium chloride and chromium sulfate have been used as dietary supplements. Chromium(III) chloride and chromium(III) sulfate can be used in the manufacture of food for particular nutritional uses and in food supplements. It is found in miscellaneous applications, such as catalysts, hardeners, mordents, crosslinking and binding agents. It is used to make very pure chromium and other chromium compounds, as a mordant in dyeing cotton, and in chromizing metals. It is too used for trivalent electroplating .
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Boiling Point | 1300°C (sublimation) |
Color | Violet |
Physical Form | Crystalline |
Quantity | 250 g |
UN Number | UN3260 |
Merck Index | 14,2222 |
Solubility Information | Partially soluble in water. |
Formula Weight | 158.35 |
Percent Purity | 98% |
Sensitivity | Hygroscopic |
Show More |
RUO – Research Use Only