missing translation for 'onlineSavingsMsg'
Learn More

Chromium(III) chloride, 99%, anhydrous

Product Code. 10752564 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Plastic bottle
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 10752564

Brand: Thermo Scientific Acros 190471000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10025-73-7 | Cl3Cr | 158.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10025-73-7
MDL Number MFCD00010948
Synonym chromium iii chloride, chromium 3+ trichloride, anhydrous chromium iii chloride, basic chromic chloride, chromium chloride, basic, chromium trichl, chromium iii ; chloride, acmc-1ai6u, ksc492k0f

Specifications

Melting Point 1152.0°C
Color Purple
Physical Form Shiny Fluffy Powder or Flakes
Quantity 100 g
Assay Percent Range 98.5% min
Molecular Formula Cl3Cr
Linear Formula CrCl3
Fieser 15,95
Merck Index 15, 2226
Solubility Information Solubility in water: slightly soluble
Molecular Weight (g/mol) 158.36
Formula Weight 158.36
Percent Purity 99%
Packaging Plastic bottle
Density 2.76
Chemical Name or Material Chromium(III) chloride, 99%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.