missing translation for 'onlineSavingsMsg'
Learn More

Chromium(III) acetate hydroxide, Cr 24%

Product Code. 11321849
Click to view available options
Quantity:
500 g
2 kg
Unit Size:
2kg
500g
This item is not returnable. View return policy

Product Code. 11321849

Brand: Thermo Scientific Alfa Aesar 012518.A3

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 39430-51-8 | Cr3(OH)2(OOCCH3)7

Occasionally to dehalogenate organic compounds. used as an oxygen scrubber, polymer industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39430-51-8
Molecular Formula Cr3(OH)2(OOCCH3)7
MDL Number MFCD00058912

Specifications

Color Blue
Physical Form Powder
Quantity 2 kg
Solubility Information Soluble in water. Insoluble in acetone. Soluble in hot water
Formula Weight 603.31
Odor Odorless
Chemical Name or Material Chromium(III) acetate hydroxide, Cr 24%

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.