missing translation for 'onlineSavingsMsg'
Learn More

Chromium(III) 2-ethylhexanoate, 50% in 2-ethylhexanoic acid

Product Code. 11487877
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
This item is not returnable. View return policy

Product Code. 11487877

Brand: Thermo Scientific Alfa Aesar A16560.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3444-17-5 | C24H45CrO6 | 481.61 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3444-17-5
Molecular Formula C24H45CrO6
Molecular Weight (g/mol) 481.61
MDL Number MFCD00014005
InChI Key WBKDDMYJLXVBNI-UHFFFAOYNA-K
Synonym chromium iii 2-ethylhexanoate, chromium 2-ethylhexanoate, hexanoic acid, 2-ethyl-, chromium 3+ salt 3:1, chromic 2-ethylhexanoate, chromium tris 2-ethylhexanoate, chromium 2-ethylhexanoate 1:3, 2-ethylhexanoic acid chromium iii salt, hexanoic acid, 2-ethyl-, chromium 3+ salt, chromium 3+ ; 2-ethylhexanoate
PubChem CID 103012
IUPAC Name chromium(3+);2-ethylhexanoate
SMILES [Cr+3].CCCCC(CC)C([O-])=O.CCCCC(CC)C([O-])=O.CCCCC(CC)C([O-])=O

Specifications

Odor Weak, Characteristic
Quantity 50 g
Formula Weight 481.62
Concentration or Composition (by Analyte or Components) 50% in 2-ethylhexanoic acid
Chemical Name or Material Chromium(III) 2-ethylhexanoate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.