missing translation for 'onlineSavingsMsg'
Learn More

Chromium(II) fluoride, 95%, Thermo Scientific™

Catalog Number 11352648
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
10049-10-2
MFCD00016044
This item is not returnable. View return policy
10049-10-2
MFCD00016044

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Boiling Point >1300°C
Melting Point 1100°C
Physical Form Powder
Quantity 5 g
UN Number UN3260
Merck Index 14,2245
Solubility Information Soluble in water,boiling hydrochloric acid. Insoluble in alcohol.
Formula Weight 89.99
Percent Purity 95%
Odor Odorless
Sensitivity Hygroscopic
Density 4.11 g/mL
Chemical Name or Material Chromium(II) fluoride
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'