missing translation for 'onlineSavingsMsg'
Learn More

Chromium(II) chloride, 90%

Product Code. p-46354 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
10 g
Packaging:
Glass bottle
Unit Size:
10g
5g
This item is not returnable. View return policy

Product Code. 10154850

Brand: Thermo Scientific Acros 223810050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10049-05-5 | Cl2Cr | 122.9 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10049-05-5
Molecular Formula Cl2Cr
Molecular Weight (g/mol) 122.9
MDL Number MFCD00010947
Synonym Chromous chloride

Specifications

Melting Point 824.0°C
Color Gray to Green
Physical Form Powder
Quantity 5 g
Assay Percent Range 88% min. (Argentometry)
Linear Formula CrCl2
Fieser 01,149; 02,76; 03,60; 04,144; 07,73; 09,511; 12,136; 13,84; 14,94; 16,93; 17,84
Merck Index 15, 2248
Solubility Information Solubility in water: may decompose. Other solubilities: insoluble in alcohol and ether
Formula Weight 122.9
Percent Purity 90%
Packaging Glass bottle
Chemical Name or Material Chromium(II) chloride, 90%
Show More Show Less
compliance-icons
Product Identifier
  • Chromium(II) chloride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.