missing translation for 'onlineSavingsMsg'
Learn More

Chromium cubes, 12.5mm (0.492in) square, 99.97% (metals basis), Thermo Scientific Chemicals

Catalog Number 11341208
Click to view available options
Quantity:
1 Ea.
5 Ea.
This item is not returnable. View return policy
7440-47-3
Cr
52.00
MFCD00010944
VYZAMTAEIAYCRO-UHFFFAOYSA-N
chrom, chrome, cromo, metal, chrome french, compounds, chrom german, chromium, elemental, chromium, metal, ccris 159
23976
CHEBI:28073
chromium
[Cr]
This item is not returnable. View return policy
7440-47-3
Cr
52.00
MFCD00010944
VYZAMTAEIAYCRO-UHFFFAOYSA-N
chrom, chrome, cromo, metal, chrome french, compounds, chrom german, chromium, elemental, chromium, metal, ccris 159
23976
CHEBI:28073
chromium
[Cr]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 1 Ea.
Percent Purity 99.97%
Odor Odorless
Weight ≈14.7g/pc
Assay (metals basis)
Chemical Name or Material Chromium cubes, 12.5mm (0.492 in.) square, Hot Isostatically Pressed

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'