missing translation for 'onlineSavingsMsg'
Learn More

Chromium carbide, 99.5% (metals basis)

Product Code. 11334368
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11334368 10 g 10g
11324368 50 g 50g
11344368 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11334368 Supplier Thermo Scientific Alfa Aesar Supplier No. 042313.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12105-81-6 | Cr23C6

Chromium carbide is useful in the surface treatment of metal components. Chromium carbide is used to coat the surface of another metal in a technique known as thermal spraying. Chromium carbide is used as an additive in cutting tools made out of cemented carbides, in order to improve toughness by preventing the growth of large grains.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12105-81-6
Molecular Formula Cr23C6
MDL Number MFCD00678358
Synonym bis methane trichromium

Specifications

Color Gray
Physical Form Powder
Quantity 10 g
Assay Percent Range 99.5% (Metal basis)
Solubility Information Reacts with water.
Formula Weight 1267.97
Odor Odorless
Appearance Gray powder
Chemical Name or Material Chromium carbide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.