missing translation for 'onlineSavingsMsg'
Learn More

Chromium boride, 99% (metals basis)

Product Code. 11337417
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11337417 25 g 25g
11347417 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11337417 Supplier Thermo Scientific Alfa Aesar Supplier No. 012570.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12007-16-8 | CrB2

Chromium boride,is been used in plasma technology and for high-performance electrodes in electron microscopes, metal matrix composites (MMCs), cutting tools and cermets for machining aluminum. chromium diboride is recognised as offering superior performance in corrosive-wear environments. Chromium diboride thin films possess desirable combinations of properties, which are attractive for a wide range of potential industrial applications. As a wear-resistant component of weld materials and powders for spraying in engineering and metallurgy, Protection of outer surfaces and bearing surfaces of machine parts against abrasive wear by plasma spraying, In aluminum evaporators manufacturing and for aluminum electrolyzes repair work, In nuclear engineering as neutron absorption material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12007-16-8
Molecular Formula CrB2
MDL Number MFCD00016043

Specifications

Color Gray
Physical Form Powder
Quantity 25 g
Assay Percent Range 99% (Metals basis)
Solubility Information It is insoluble in water.
Formula Weight 73.62
Odor Odorless
Chemical Name or Material Chromium boride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.