Learn More
Manutan Chrome wire shelving extension bay, 1880mm x 1220mm x 355mm
Description
To create long runs of shelving. Adjustable, simple to assemble and easy to clean. Suitable for use in food and medical areas where a high standard of hygiene must be maintained. Supplied with shelf hooks to attach additional bays. Self assembly - No nuts and bolts required.
- Four shelves
- Shelf capacity UDL: 175kg
- Dimensions (H x W x D): 1880mm x 1220mm x 355mm
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.