missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Manutan Chrome shelf, 1520mm x 455mm, pack of two
Brand: Manutan A362390
Code : Z3
Additional Details : Weight : 1.00000kg
Description
Strong wire mesh shelves for extra storage space with triple nickel and chrome plated finish. Suitable for use in food and medical areas where a high standard of hygiene must be maintained. Supplied in packs of two with support clips.Dimensions (W x D): 1520mm x 455mm