Learn More
Pro-Lab Diagnostics™ Nichrome Inoculation Loops
Calibrated to remove a consistent, precise amount of material. Pro-Lab Diagnostics™ Nichrome Inoculation Loops are economical nickel-chrome wire loops in a range of sizes from 1μL to 10μL. The standard and heavy gauge wire loops can be used with insulated holders (sold separately).
Brand:  Pro-Lab Diagnostics™ PL-103
Code : 04
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 12830155
  • £41.95 / Pack of 25
Estimated Shipment: 03-05-2024
to see stock.
Add to basket
Description
Description
An inoculation loop is a simple tool used to gather and transfer microscopic organisms from a culture or broth plate. The small loop removes a consistent amount of inoculum by using surface tension. The inoculation loop is flame-sterilized before and after each use to eliminate cross-contamination between cultures. (The loop must be cooled after sterilization to avoid destroying the organisms.)
  • Constructed of economical nickel-chrome in standard or heavy gauge wire
  • Available in a range of 6 sizes, 1μL / 1.59mmD to 10μL / 5.05mmD
  • Packaged in sets of 25 loops
  • Holders with insulated handles sold separately
Specifications
Specifications
2.52 mm
2.5 μL
Standard
Yes
Chrome-Nickel
Inoculating Loop
25/Pk.