missing translation for 'onlineSavingsMsg'
Learn More

Chrome Azurol S, MP Biomedicals™

Product Code. 11493182 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Unit Size:
100g
10g
25g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11493182 10 g 10g
11427554 10 g 10g
11403192 25 g 25g
11413192 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11493182 Supplier MP Biomedicals Supplier No. 0215498210

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chrome Azurol S is used as a reagent for determination of fluoride

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1667-99-8
Molecular Formula C23H13Cl2Na3O9S
Molecular Weight (g/mol) 605.277
MDL Number MFCD00001615
InChI Key FUIZKNBTOOKONL-UHFFFAOYSA-K
Synonym Mordant Blue 29, C.I. 43825
PubChem CID 54736314
IUPAC Name trisodium;5-[(3-carboxy-5-methyl-4-oxocyclohexa-2,5-dien-1-ylidene)-(2,6-dichloro-3-sulfonatophenyl)methyl]-3-methyl-2-oxidobenzoate
SMILES CC1=CC(=CC(=C1[O-])C(=O)[O-])C(=C2C=C(C(=O)C(=C2)C(=O)O)C)C3=C(C=CC(=C3Cl)S(=O)(=O)[O-])Cl.[Na+].[Na+].[Na+]

Specifications

Formula Weight 605.29
Color Brown
Quantity 10 g
Physical Form Powder
Chemical Name or Material Chrome Azurol S, Dye content: approx. 65%
compliance-icons
Product Identifier
  • Chrome Azurol S
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Chrome Azurol S
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.