missing translation for 'onlineSavingsMsg'
Learn More

Hach Lange™ ChromaVer™ 3 Chromium Reagent Powder Pillows

Product Code. 15918100
Change view
Click to view available options
Quantity:
100/Pk.
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15918100 100/Pk. Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15918100 Supplier Hach Lange™ Supplier No. 1271099

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For determination of hexavalent chromium by the Diphenylcarbohydrazide method.

For determination of hexavalent chromium by the Diphenylcarbohydrazide method.USEPA accepted for reporting for wastewater analyses using Hach Method 8023, Diphenylcarbohydrazide Method.Photometric Sample Size: 10 mL. Photometric Range: up to 0.70 mg/L.Kit Sample Size: 5 mL. Kit Range: Up to 1.50 Pack of 100 powder pillows.

    To confirm accurate results, see appropriate standard solutions on the Consumables tab.

    Specifications

  • EPA compliant: Yes
  • Instrument: All except DR/3000, DR/2000, DR 900
  • Method: 8023
  • Method Name: 1,5-Diphenylcarbohydrazide
  • Number of tests: 100
  • Parameter: Chromium
  • Platform: Powder Pillows
  • Quantity: /pk
  • Range: 0.010 - 0.700 mg/L Cr⁶⁺

Specifications

Volume (Metric) 5 or 10 mL
Quantity 100/Pk.
Range 0.010 to 0.700 mg/L Cr6+
Type Powder Pillows
compliance-icons
Product Identifier
  • 12710-99 ChromaVer 3
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Potassium disulfate,Magnesium sulfate,1,5 Diphenylcarbazide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.