missing translation for 'onlineSavingsMsg'
Learn More

ROBU VitraPOR™ Chromatography-Column

Product Code. 16883086
Click to view available options
Column Capacity:
20 mL
85 mL
Pore Size:
1
2
3
4
Unit Size:
Pack of 5
This item is not returnable. View return policy

Product Code. 16883086

Brand: ROBU VitraPOR™ 31104

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Chromatography-Column with fused filter disc.

  • Highly resistant to most chemicals and temperatures up to 500°C

Specifications

Quantity 5/Pk.
Pore Size 4
Column Capacity 20 mL
Material Borosilicate Glass 3.3
Includes Fused Filter Disc
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.