missing translation for 'onlineSavingsMsg'
Learn More

(Chroman-8-ylmethyl)amine, 90%, Thermo Scientific™

Product Code. 10381710 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10381710 5 g Glass bottle 5g
10422980 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10381710 Supplier Thermo Scientific Acros Supplier No. 432660050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 933727-40-3
Molecular Weight (g/mol) 163.22
MDL Number MFCD17171385
InChI Key CPJFACCHHWIMEM-UHFFFAOYSA-N
Synonym chroman-8-ylmethyl amine, 2h-1-benzopyran-8-methanamine, 3,4-dihydro, 1-3,4-dihydro-2h-1-benzopyran-8-yl methanamine
PubChem CID 82596485
IUPAC Name 3,4-dihydro-2H-chromen-8-ylmethanamine
SMILES C1CC2=C(C(=CC=C2)CN)OC1

Specifications

CAS Min % 88.0
CAS Max % 100.0
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 88% min. (GC)
Packaging Glass bottle
Molecular Formula C10H13NO
Quantity 5 g
Formula Weight 163.22
Percent Purity 90%
Physical Form Liquid
Chemical Name or Material (Chroman-8-ylmethyl)amine
Show More Show Less
compliance-icons
Product Identifier
  • (Chroman-8-ylmethyl)amine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.