missing translation for 'onlineSavingsMsg'
Learn More

Chroman-3-carboxylic acid, 97%, Thermo Scientific Chemicals

Product Code. 15404738
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15404738 1 g 1g
15414738 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15404738 Supplier Thermo Scientific Alfa Aesar Supplier No. H58448.03
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 115822-57-6 | Molecular Formula: C10H10O3 | Molecular Weight: 178.187 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 115822-57-6
Molecular Formula C10H10O3
Molecular Weight (g/mol) 178.187
MDL Number MFCD03783554
InChI Key UGAGZMGJJFSKQM-UHFFFAOYSA-N
Synonym chroman-3-carboxylic acid, 3-chromanecarboxylic acid, chromane-3-carboxylic acid, 3,4-dihydro-2h-1-benzopyran-3-carboxylic acid, 2h-1-benzopyran-3-carboxylic acid, 3,4-dihydro, 2h-1-benzopyran-3-carboxylicacid, 3,4-dihydro, 3,4-dihydro-2h-benzopyran-3-carboxylic acid
PubChem CID 2795458
IUPAC Name 3,4-dihydro-2H-chromene-3-carboxylic acid
SMILES C1C(COC2=CC=CC=C21)C(=O)O

Specifications

Melting Point 117°C to 121°C
Quantity 1 g
Formula Weight 178.18
Percent Purity 97%
Chemical Name or Material Chroman-3-carboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.